3-Aminobenzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR1944-100G OR1944-500G
APOSOR1944-100GEA 0 EUR
APOSOR1944-100G APOSOR1944-500G
3-Aminobenzoic acid 97%
3-Aminobenzoic acid
Formule: C₇H₇NO₂
Smeltpunt: 172…180 °C
Dichtheid: 1,506 g/cm³ (25 °C)
Vlampunt: 150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007795
CAS nummer: 99-05-8
EINECS: 202-724-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR