5-Bromo-4-chloro-2-aminopyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Amino-5-bromo-4-chloropyridine, 5-Bromo-4-chloropyridin-2-amine

OR19535-1G OR19535-5G
APOSOR19535-1GEA 22 EUR
APOSOR19535-1G APOSOR19535-5G
5-Bromo-4-chloro-2-aminopyridine 97%
5-Bromo-4-chloro-2-aminopyridine
Formule: C₅H₄BrClN₂
Smeltpunt: 141…147 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12407282
CAS nummer: 942947-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR