4′-Azido-2-bromoacetophenone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR2000T-1G OR2000T-250MG
APOSOR2000T-1GEA 584 EUR
APOSOR2000T-1G APOSOR2000T-250MG
4′-Azido-2-bromoacetophenone
4′-Azido-2-bromoacetophenone

A versatile photolabile bifunctional reagent for investigating the active sites of sulphydryl enzymes,particularly those which poses a reactive cysteine residue at the active site.

Formule: C₈H₆BrN₃O
Smeltpunt: 68…70 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00042637
CAS nummer: 57018-46-9
EINECS: 260-519-5
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR