Bromotrimethylsilane

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR2120-100G OR2120-500G OR2120-25G
APOSOR2120-100GEA 38 EUR
APOSOR2120-100G APOSOR2120-500G APOSOR2120-25G
Bromotrimethylsilane
Bromotrimethylsilane
Formule: C₃H₉BrSi
Kookpunt: 79 °C
Smeltpunt: –43 °C
Dichtheid: 1,17 g/cm³ (25 °C)
Vlampunt: –8 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000048
CAS nummer: 2857-97-8
EINECS: 220-672-0
UN: 2924
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR