3-(3-Nitrophenyl)-1,2,4-oxadiazole
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR21205-1G
OR21205-250MG
APOSOR21205-1GEA
330
EUR
APOSOR21205-1G
APOSOR21205-250MG
3-(3-Nitrophenyl)-1,2,4-oxadiazole
3-(3-Nitrophenyl)-1,2,4-oxadiazole
Formule:
C₈H₅N₃O₃ Bewaar temperatuur: Kamertemperatuur |
CAS nummer:
90049-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...