6-Chloroanthranilic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Amino-6-chlorobenzoic acid

OR21294-100G OR21294-25G OR21294-5G
APOSOR21294-100GEA 36.6 EUR
APOSOR21294-100G APOSOR21294-25G APOSOR21294-5G
6-Chloroanthranilic acid
6-Chloroanthranilic acid
Formule: C₇H₆ClNO₂
Smeltpunt: 142…145 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051530
CAS nummer: 2148-56-3
EINECS: 218-416-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR