4-Ethoxyphenylacetylene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-Ethoxy-4-ethynylbenzene, 4-Ethoxyphenylacetylene

OR21950-100G OR21950-25G
APOSOR21950-100GEA 336 EUR
APOSOR21950-100G APOSOR21950-25G
4-Ethoxyphenylacetylene 98%
4-Ethoxyphenylacetylene
Formule: C₁₀H₁₀O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00173880
CAS nummer: 79887-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR