2-Amino-N-cyclohexylbenzamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR2218-1G OR2218-250MG OR2218-5G
APOSOR2218-1GEA 58.4 EUR
APOSOR2218-1G APOSOR2218-250MG APOSOR2218-5G
2-Amino-N-cyclohexylbenzamide
2-Amino-N-cyclohexylbenzamide
Formule: C₁₃H₁₈N₂O
Smeltpunt: 153…156 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00060339
CAS nummer: 56814-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR