5,6-Diaminouracil hydrochloride
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 5,6-Diamino-2,4-dihydroxypyrimidine hydrochloride, 5,6-Diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
OR22288-1G OR22288-5G
APOSOR22288-1GEA 103 EUR
APOSOR22288-1G APOSOR22288-5G
5,6-Diaminouracil hydrochloride
5,6-Diaminouracil hydrochloride
Formule: C₄H₆N₄O₂.HCl
MW: 178.58 g/mol
MDL Nummer: MFCD00035307
CAS nummer: 53608-89-2