3-Iodophthalic acid 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR22495-25G
OR22495-100G
APOSOR22495-25GEA
95
EUR
APOSOR22495-25G
APOSOR22495-100G
3-Iodophthalic acid 95%
3-Iodophthalic acid
Formule:
C₈H₅IO₄ Smeltpunt: 178…182 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00275568 CAS nummer: 6937-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...