N-Cbz-4-piperidinecarboxylic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR23101-5G OR23101-1G
APOSOR23101-5GEA 0 EUR
APOSOR23101-5G APOSOR23101-1G
N-Cbz-4-piperidinecarboxylic acid 97%
N-Cbz-4-piperidinecarboxylic acid
Formule: C₁₄H₁₇NO₄
Smeltpunt: 76…81 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01568759
CAS nummer: 10314-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR