4-(Bromomethyl)-5-phenyloxazole 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR23139-250MG
OR23139-100MG
APOSOR23139-250MGEA
0
EUR
APOSOR23139-250MG
APOSOR23139-100MG
4-(Bromomethyl)-5-phenyloxazole 95%
4-(Bromomethyl)-5-phenyloxazole
Formule:
C₁₀H₈BrNO Smeltpunt: 50…52 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03407316 CAS nummer: 368869-94-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...