Pivaloylacetonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile

OR23696-100G OR23696-250G OR23696-25G
APOSOR23696-100GEA 35.1 EUR
APOSOR23696-100G APOSOR23696-250G APOSOR23696-25G
Pivaloylacetonitrile
Pivaloylacetonitrile
Formule: C₇H₁₁NO
Kookpunt: 125…126 °C/15mmHg
Smeltpunt: 66…70 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010208
CAS nummer: 59997-51-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR