3-Methylenecyclobutanecarbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR2438-25G OR2438-5G
APOSOR2438-25GEA 151 EUR
APOSOR2438-25G APOSOR2438-5G
3-Methylenecyclobutanecarbonitrile
3-Methylenecyclobutanecarbonitrile
Formule: C₆H₇N
Kookpunt: 149…150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01085401
CAS nummer: 15760-35-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR