Phthalan 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR24902-100G OR24902-25G OR24902-5G
APOSOR24902-100GEA 337 EUR
APOSOR24902-100G APOSOR24902-25G APOSOR24902-5G
Phthalan 98%
Phthalan
Formule: C₈H₈O
Kookpunt: 190…192 °C
Dichtheid: 1,098 g/cm³ (25 °C)
Vlampunt: 63 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005932
CAS nummer: 496-14-0
EINECS: 207-815-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR