3-Ethynylaniline 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR2625-25G OR2625-5G OR2625-100G
APOSOR2625-25GEA 30.8 EUR
APOSOR2625-25G APOSOR2625-5G APOSOR2625-100G
3-Ethynylaniline 99%
3-Ethynylaniline
Formule: C₈H₇N
Kookpunt: 92…93 °C/2mmHg
Dichtheid: 1,12 g/cm³ (25 °C)
Vlampunt: 59 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00014779
CAS nummer: 54060-30-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR