Ethyl acetopyruvate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR27077-25G OR27077-5G
APOSOR27077-25GEA 71.6 EUR
APOSOR27077-25G APOSOR27077-5G
Ethyl acetopyruvate 97%
Ethyl acetopyruvate
Formule: C₇H₁₀O₄
Kookpunt: 101…103 °C/12mmHg
Smeltpunt: 16…18 °C
Dichtheid: 1,126 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009124
CAS nummer: 615-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR