1-Adamantanecarbonitrile
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR27249-5G
OR27249-250MG
APOSOR27249-5GEA
103
EUR
APOSOR27249-5G
APOSOR27249-250MG
1-Adamantanecarbonitrile
1-Adamantanecarbonitrile
Formule:
C₁₁H₁₅N Smeltpunt: 193…196 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00074731 CAS nummer: 23074-42-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...