2-Chloro-3-nitropyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR28619-100G OR28619-25G
APOSOR28619-100GEA 95 EUR
APOSOR28619-100G APOSOR28619-25G
2-Chloro-3-nitropyridine
2-Chloro-3-nitropyridine
Formule: C₅H₃ClN₂O₂
Smeltpunt: 100…103 °C
Vlampunt: 185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006232
CAS nummer: 5470-18-8
EINECS: 226-799-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR