Cyclobutanecarbonyl chloride 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR28716-100G
OR28716-25G
APOSOR28716-100GEA
310
EUR
APOSOR28716-100G
APOSOR28716-25G
Cyclobutanecarbonyl chloride 95%
Cyclobutanecarbonyl chloride
Formule:
C₅H₇ClO Kookpunt: 60 °C/50mmHg Dichtheid: 1,039 g/cm³ (25 °C) Vlampunt: 37 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001319 CAS nummer: 5006-22-4 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...