4,4'-Bipyridyl 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR28750-25G OR28750-5G
APOSOR28750-25GEA 35.1 EUR
APOSOR28750-25G APOSOR28750-5G
4,4'-Bipyridyl 97%
4,4'-Bipyridyl
Formule: C₁₀H₈N₂
Kookpunt: 304…305 °C
Smeltpunt: 109…114 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006416
CAS nummer: 553-26-4
EINECS: 209-036-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR