3-tert-Butyl-1H-pyrazole-5-carbohydrazide
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR29615-250MG
OR29615-1G
APOSOR29615-250MGEA
58.4
EUR
APOSOR29615-250MG
APOSOR29615-1G
3-tert-Butyl-1H-pyrazole-5-carbohydrazide
3-tert-Butyl-1H-pyrazole-5-carbohydrazide
Formule:
C₈H₁₄N₄O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00619318 CAS nummer: 262292-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...