4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR301166-1G OR301166-250MG
APOSOR301166-1GEA 0 EUR
APOSOR301166-1G APOSOR301166-250MG
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine
Formule: C₈H₇BrN₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD15529172
CAS nummer: 1014613-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR