4-Bromophthalimide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR302268-100G OR302268-25G
APOSOR302268-100GEA 158 EUR
APOSOR302268-100G APOSOR302268-25G
4-Bromophthalimide
4-Bromophthalimide
Formule: C₈H₄BrNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00466049
CAS nummer: 6941-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR