Methyl-6-piperazinonicotinate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR303741-1G OR303741-100MG OR303741-250MG
APOSOR303741-1GEA 262 EUR
APOSOR303741-1G APOSOR303741-100MG APOSOR303741-250MG
Methyl-6-piperazinonicotinate
Methyl-6-piperazinonicotinate
Formule: C₁₁H₁₅N₃O₂
Smeltpunt: 165…167 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00894966
CAS nummer: 132144-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR