Methyl-4-(pyrrolidin-1-ylmethyl)benzoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR303935-1G OR303935-500MG
APOSOR303935-1GEA 0 EUR
APOSOR303935-1G APOSOR303935-500MG
Methyl-4-(pyrrolidin-1-ylmethyl)benzoate
Methyl-4-(pyrrolidin-1-ylmethyl)benzoate
Formule: C₁₃H₁₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01452504
CAS nummer: 160598-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR