4-Methoxyresorcinol
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR305005-1G
OR305005-250MG
APOSOR305005-1GEA
0
EUR
APOSOR305005-1G
APOSOR305005-250MG
4-Methoxyresorcinol
4-Methoxyresorcinol
Formule:
C₇H₈O₃ Kookpunt: 115…116 °C/1mmHg Smeltpunt: 66…72 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00210443 CAS nummer: 6100-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...