1-Methyl-1H-indazole-6-carboxylic acid methyl ester
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR30720-1G
OR30720-250MG
APOSOR30720-1GEA
26.3
EUR
APOSOR30720-1G
APOSOR30720-250MG
1-Methyl-1H-indazole-6-carboxylic acid methyl ester
1-Methyl-1H-indazole-6-carboxylic acid methyl ester
Formule:
C₁₀H₁₀N₂O₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD11109403 CAS nummer: 1007219-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...