3-Amino-1-benzylpiperidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR30968-1G OR30968-5G OR30968-25G
APOSOR30968-1GEA 38.9 EUR
APOSOR30968-1G APOSOR30968-5G APOSOR30968-25G
3-Amino-1-benzylpiperidine
3-Amino-1-benzylpiperidine
Formule: C₁₂H₁₈N₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD04038444
CAS nummer: 60407-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR