D(+)-Tryptophan 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
H-D-Trp-OH
OR315019-100G
OR315019-500G
APOSOR315019-100GEA
71.6
EUR
APOSOR315019-100G
APOSOR315019-500G
D(+)-Tryptophan 97%
D-(+)-Tryptophan
Formule:
C₁₁H₁₂N₂O₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005647 CAS nummer: 153-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...