1-(5-Bromo-2-furyl)ethanone
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-(5-Bromofuran-2-yl)ethanone
OR315519-1G
OR315519-250MG
OR315519-500MG
APOSOR315519-1GEA
706
EUR
APOSOR315519-1G
APOSOR315519-250MG
APOSOR315519-500MG
1-(5-Bromo-2-furyl)ethanone
1-(5-Bromo-2-furyl)ethanone
Formule:
C₆H₅BrO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD06657513 CAS nummer: 3199-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...