4-Pentylcyclohexanecarbohydrazide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR33412-1G OR33412-5G
APOSOR33412-1GEA 541 EUR
APOSOR33412-1G APOSOR33412-5G
4-Pentylcyclohexanecarbohydrazide 95%
4-Pentylcyclohexanecarbohydrazide
Formule: C₁₂H₂₄N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03425827
CAS nummer: 454473-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR