Indole-6-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR3398-100G OR3398-25G OR3398-5G
APOSOR3398-100GEA 134 EUR
APOSOR3398-100G APOSOR3398-25G APOSOR3398-5G
Indole-6-carboxylic acid
Indole-6-carboxylic acid
Formule: C₉H₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00210441
CAS nummer: 1670-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR