Smart Station ultrapure water remote dispenser, Arium®
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H2O-ARST-UP-T H2O-ARST-UP-B
171-1396EA 2290 EUR
171-1396 171-1395
Smart Station ultrapure water remote dispenser, Arium®
Waterdispensers
The Arium® Smart Station can be optimised to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispense pure or ultrapure water. The Smart Station ultrapure supports multiple points of use to save costs and valuable lab space.
  • Compact: Save space integrating into any lab
  • Intuitive: Touch-activated colour display with direct access to all important functions
  • Flexible: Stepless height adjustment to fill different size containers
  • Ergonomic design: Hand-held for optimised left- or right-hand operation
  • Accurate: Precise volume dispensing for reliable buffer and sample preparation
Flexible remote dispenser for Arium® Pro and Comfort systems. Height adjustable display/dispense station for dispensing Type 1 ultrapure water.
Intended for use with Arium®Pro DI, Pro UF, Pro UV, Pro VF, and Arium® Comfort.
Leveringsinformatie: Equipment supplied: Display/dispense unit with stand, power supply including IEC connector, connection set.