Smart Station pure water remote dispenser, Arium®
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
arium®
171-1393EA 1552 EUR
171-1393 171-1394
Smart Station pure water remote dispenser, Arium®
Waterdispensers
The Arium® Smart Station can be optimised to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispense pure water. Smart Station pure supports multiple points of use to save costs and valuable lab space.
  • Compact: Save space integrating into any lab
  • Intuitive: Touch-activated colour display with direct access to all important functions
  • Flexible: Stepless height adjustment to fill different size containers
  • Ergonomic design: Hand-held for optimised left- or right-hand operation
  • Accurate: Precise volume dispensing for reliable buffer and sample preparation
Flexible remote dispenser for Arium® Bagtank. Height adjustable display/dispense station for dispensing pure water.
Leveringsinformatie: Equipment supplied: Display/dispense unit with stand, power supply including IEC connector, connection set.