4-Chloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR350090-1G OR350090-250MG OR350090-5G
APOSOR350090-1GEA 220 EUR
APOSOR350090-1G APOSOR350090-250MG APOSOR350090-5G
4-Chloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine
4-Chloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine
Formule: C₇H₇ClN₂
MDL Nummer: MFCD12755922
CAS nummer: 83942-13-6