Isoxazole-4-boronic acid 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR360562-250MG
OR360562-1G
OR360562-5G
APOSOR360562-250MGEA
0
EUR
APOSOR360562-250MG
APOSOR360562-1G
APOSOR360562-5G
Isoxazole-4-boronic acid 98%
Isoxazole-4-boronic acid
Formule:
C₃H₄BNO₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD06657892 CAS nummer: 1008139-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...