6-Aminophthalide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR3916-1G OR3916-5G
APOSOR3916-1GEA 0 EUR
APOSOR3916-1G APOSOR3916-5G
6-Aminophthalide 95%
6-Aminophthalide
Formule: C₈H₇NO₂
Smeltpunt: 172…175 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00033530
CAS nummer: 57319-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR