2-Amino-5-ethoxypyrazine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR40077-1G OR40077-250MG
APOSOR40077-1GEA 342 EUR
APOSOR40077-1G APOSOR40077-250MG
2-Amino-5-ethoxypyrazine
2-Amino-5-ethoxypyrazine
Formule: C₆H₉N₃O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09909690
CAS nummer: 647843-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR