1-(1-Ethylpropyl)piperazine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR4170-1G OR4170-5G
APOSOR4170-1GEA 58.4 EUR
APOSOR4170-1G APOSOR4170-5G
1-(1-Ethylpropyl)piperazine
1-(1-Ethylpropyl)piperazine
Formule: C₉H₂₀N₂
Kookpunt: 86 °C/11,2mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02093554
CAS nummer: 373356-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR