1-Iodo-4-nitrobenzene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR4306-100G OR4306-10G
APOSOR4306-100GEA 0 EUR
APOSOR4306-100G APOSOR4306-10G
1-Iodo-4-nitrobenzene 98%
1-Iodo-4-nitrobenzene
Formule: C₆H₄INO₂
Kookpunt: 289 °C/772mmHg
Smeltpunt: 173 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007299
CAS nummer: 636-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR