5-Nitro-1,2,4-trichlorobenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR4439-25G OR4439-500G
APOSOR4439-25GEA 0 EUR
APOSOR4439-25G APOSOR4439-500G
5-Nitro-1,2,4-trichlorobenzene
5-Nitro-1,2,4-trichlorobenzene
Formule: C₆H₂Cl₃NO₂
Kookpunt: 150 °C/15mmHg
Smeltpunt: 58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007072
CAS nummer: 89-69-0
EINECS: 201-931-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR