tert-Butyl-2-piperidinecarboxylate 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR452116-1G
OR452116-500MG
APOSOR452116-1GEA
526
EUR
APOSOR452116-1G
APOSOR452116-500MG
tert-Butyl-2-piperidinecarboxylate 97%
tert-Butyl-2-piperidinecarboxylate
Formule:
C₁₀H₁₉NO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD11867900 CAS nummer: 147202-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...