β-Methyllevulinic acid
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR46668-1G
OR46668-250MG
OR46668-500MG
OR46668-5G
APOSOR46668-1GEA
220
EUR
APOSOR46668-1G
APOSOR46668-250MG
APOSOR46668-500MG
APOSOR46668-5G
β-Methyllevulinic acid
beta-Methyllevulinic acid
Formule:
C₆H₁₀O₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00143691 CAS nummer: 6628-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...