Benzyltriethylammonium chloride 99%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Benzyl triethylammonium chloride
OR4667-500G OR4667-5KG OR4667-1KG
APOSOR4667-500GEA 24.9 EUR
APOSOR4667-500G APOSOR4667-5KG APOSOR4667-1KG
Benzyltriethylammonium chloride 99%
Benzyltriethylammonium chloride
Formule: C₁₃H₂₂ClN
MW: 227.78 g/mol
Kookpunt: 444.8 °C
Smeltpunt: 185…192 °C (ontbinding)
Vlampunt: >275 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011824
CAS nummer: 56-37-1
EINECS: 200-270-1