3-Acetylquinoline
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR470156-1G
OR470156-5G
APOSOR470156-1GEA
64.3
EUR
APOSOR470156-1G
APOSOR470156-5G
3-Acetylquinoline
3-Acetylquinoline
Formule:
C₁₁H₉NO Kookpunt: 139 °C Smeltpunt: 97…99 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00030698 CAS nummer: 33021-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...