4-Iodoisatin
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR471590-1G
OR471590-5G
APOSOR471590-1GEA
42.4
EUR
APOSOR471590-1G
APOSOR471590-5G
4-Iodoisatin
4-Iodoisatin
Formule:
C₈H₄INO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03427300 CAS nummer: 20780-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...