Iodocyclopentane 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR480594-50G OR480594-100G
APOSOR480594-50GEA 205 EUR
APOSOR480594-50G APOSOR480594-100G
Iodocyclopentane 97%
Iodocyclopentane
Formule: C₅H₉I
MDL Nummer: MFCD00039446
CAS nummer: 1556-18-9