4-Isopropylbenzenesulphonamide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR480691-25G OR480691-5G
APOSOR480691-25GEA 704 EUR
APOSOR480691-25G APOSOR480691-5G
4-Isopropylbenzenesulphonamide 97%
4-Isopropylbenzenesulphonamide
Formule: C₉H₁₃NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00457071
CAS nummer: 6335-39-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR