3-Nitro-p-toluonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Methyl-3-nitrobenzonitrile

OR4834-25G OR4834-5G
APOSOR4834-25GEA 0 EUR
APOSOR4834-25G APOSOR4834-5G
3-Nitro-p-toluonitrile
3-Nitro-p-toluonitrile
Formule: C₈H₆N₂O₂
Kookpunt: 171 °C/12mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00031482
CAS nummer: 939-79-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR